Assay Details
Gene Symbol: Gucd1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
GenBank mRNA | BC169010.1 | - | 161 | 9098752 | 85 | |
KC686830.1 | - | 161 | 85 |

More Information
Additional Information:
This assay is in the 5'UTR region of the BC169010.1, KC686830.1 transcripts.
Set Membership: |
![]() ![]() ![]() ![]() |
MicroArray Probes:
