Assay Details
Gene Symbol: Tap1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_032055.4 | NP_114444.2 | 3 - 4 | 837 | 126 | |
GenBank mRNA | BC101854.1 | - | 3 - 4 | 1104 | 126 | |
KC222902.1 | - | 3 - 4 | 778 | 126 | ||
KC222904.1 | - | 3 - 4 | 778 | 126 | ||
KC222905.1 | - | 3 - 4 | 595 | 126 | ||
KC222906.1 | - | 3 - 4 | 778 | 126 | ||
X57523.1 | - | 3 - 4 | 825 | 126 | ||
Y10230.1 | - | 3 - 4 | 778 | 126 | ||
Y10231.1 | - | 3 - 4 | 778 | 126 | ||
Y10232.1 | - | 3 - 4 | 778 | 126 | ||
Y10233.1 | - | 3 - 4 | 778 | 126 | ||
Y10234.1 | - | 3 - 4 | 778 | 126 | ||
Y10235.1 | - | 3 - 4 | 778 | 126 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
