Assay Details
Gene Symbol: Mapk14 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_031020.2 | NP_112282.2 | 3 - 4 | 606 | 102 | |
XM_006256142.2 | XP_006256204.1 | 3 - 4 | 318 | 102 | ||
GenBank mRNA | AB000783.1 | - | 3 - 4 | 605 | 102 | |
AF346293.1 | - | 3 - 4 | 303 | 102 | ||
BC092193.1 | - | 3 - 4 | 606 | 7313217 | 102 | |
U91847.1 | - | 3 - 4 | 303 | 102 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
