Assay Details
Gene Symbol: Tnfaip2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001137633.1 | NP_001131105.1 | 11 - 12 | 2096 | 59 | |
XM_006240577.2 | XP_006240639.1 | 10 - 11 | 2104 | 59 | ||
GenBank mRNA | FQ212109.1 | - | 10 - 11 | 1854 | 59 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() |
Gene Ontology Categories:
MicroArray Probes:
