Assay Details
Gene Symbol: Tnfaip2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001137633.1 | NP_001131105.1 | 3 - 4 | 621 | 60 | |
XM_006240577.2 | XP_006240639.1 | 2 - 3 | 629 | 60 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() |
Gene Ontology Categories:
MicroArray Probes:
