Assay Details
Gene Symbol: Tnfaip2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001137633.1 | NP_001131105.1 | 8 - 9 | 1688 | 65 | |
XM_006240577.2 | XP_006240639.1 | 7 - 8 | 1696 | 65 | ||
GenBank mRNA | FQ212109.1 | - | 7 - 8 | 1446 | 65 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() |
Gene Ontology Categories:
MicroArray Probes:
