Assay Details
Gene Symbol: Crip2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_008764943.1 | XP_008763165.1 | 1 - 2 | 364 | 178 |

More Information
Additional Information:
This assay is in the 5'UTR region of the XM_008764943.1 transcripts.
Set Membership: |
![]() ![]() ![]() ![]() |
Gene Ontology Categories:
