Assay Details
Gene Symbol: Wars | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001013170.2 | NP_001013188.2 | 1 - 2 | 59 | 61 | |
XM_006240542.2 | XP_006240604.1 | 1 - 2 | 96 | 61 | ||
GenBank mRNA | AY724504.1 | - | 1 - 2 | 59 | 61 | |
BC061752.1 | - | 1 - 2 | 55 | 5600013 | 61 |

More Information
Additional Information:
This assay is in the 5'UTR region of the BC061752.1, NM_001013170.2, XM_006240542.2 transcripts.
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
