Assay Details
Gene Symbol: Wars | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001013170.2 | NP_001013188.2 | 2 - 3 | 223 | 105 | |
XM_006240542.2 | XP_006240604.1 | 2 - 3 | 260 | 105 | ||
XM_008764811.1 | XP_008763033.1 | 2 - 3 | 414 | 105 | ||
XM_008764812.1 | XP_008763034.1 | 2 - 3 | 1011 | 105 | ||
GenBank mRNA | AY724504.1 | - | 2 - 3 | 223 | 105 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
