Assay Details
Gene Symbol: Wars | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001013170.2 | NP_001013188.2 | 8 - 9 | 1066 | 62 | |
XM_006240542.2 | XP_006240604.1 | 8 - 9 | 1103 | 62 | ||
XM_008764811.1 | XP_008763033.1 | 8 - 9 | 1257 | 62 | ||
XM_008764812.1 | XP_008763034.1 | 8 - 9 | 1854 | 62 | ||
GenBank mRNA | AY724504.1 | - | 8 - 9 | 1066 | 62 | |
BC061752.1 | - | 9 - 10 | 1129 | 5600013 | 62 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
