Assay Details
Gene Symbol: Wdr20 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_006240583.2 | XP_006240645.1 | 3 - 4 | 1761 | 72 | |
XM_006240585.2 | XP_006240647.1 | 2 - 3 | 1578 | 72 | ||
XM_008764933.1 | XP_008763155.1 | 4 - 5 | 1955 | 72 | ||
XM_008764934.1 | XP_008763156.1 | 4 - 5 | 1904 | 72 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
MicroArray Probes:
