Assay Details
Gene Symbol: Bdkrb2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001270713.1 | NP_001257642.1 | 1 - 2 | 56 | 89 | |
NM_173100.2 | NP_775123.2 | 1 - 2 | 99 | 89 | ||
GenBank mRNA | M59967.1 | - | 1 - 2 | 56 | 89 | |
X69681.1 | - | 1 - 2 | 99 | 89 |

More Information
Additional Information:
This assay is in the 5'UTR region of the M59967.1, NM_001270713.1, NM_173100.2, X69681.1 transcripts.
Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
