Assay Details
Gene Symbol: Bdkrb2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001270713.1 | NP_001257642.1 | 2 - 3 | 160 | 131 | |
GenBank mRNA | M59967.1 | - | 2 - 3 | 160 | 131 |

More Information
Updated Mapping Information

Additional Information:
This assay is in the 5'UTR region of the M59967.1, NM_001270713.1 transcripts.
Set Membership: |
![]() ![]() ![]() ![]() |
Gene Ontology Categories:
MicroArray Probes:
