Assay Details
Gene Symbol: Adssl1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_003750237.3 | XP_003750285.1 | 11 - 12 | 1252 | 67 | |
XM_006240653.2 | XP_006240715.1 | 11 - 12 | 1245 | 67 |

More Information
Set Membership: |
![]() ![]() ![]() |
Gene Ontology Categories:
