Assay Details
Gene Symbol: Mok | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001010965.1 | NP_001010965.1 | 5 - 6 | 508 | 116 | |
XM_006240589.2 | XP_006240651.2 | 5 - 6 | 668 | 116 | ||
XM_006240590.2 | XP_006240652.2 | 5 - 6 | 668 | 116 | ||
XM_006240593.2 | XP_006240655.2 | 5 - 6 | 668 | 116 | ||
XM_008764945.1 | XP_008763167.1 | 2 - 3 | 392 | 116 | ||
GenBank mRNA | BC079440.1 | - | 5 - 6 | 508 | 116 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
