Assay Details
Gene Symbol: Cnksr3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001012061.1 | NP_001012061.1 | 12 - 13 | 1884 | 93 | |
XM_006227875.2 | XP_006227937.1 | 12 - 13 | 3041 | 93 | ||
GenBank mRNA | AY328890.1 | - | 12 - 13 | 1884 | 93 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
Agilent
Affymetrix
ABI
Illumina
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Agilent Whole Rat Genome Microarray 4x44K v2 | A_64_P158148 | - | - |
Agilent Whole Rat Genome Microarray 4x44K v2 | A_44_P182402 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 791538 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 965780 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 667996 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 31316 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 864217 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 690815 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 445974 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 741569 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 449441 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 902177 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 661401 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 891811 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 363537 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 94496 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 636227 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 535274 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 364516 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 146264 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 517354 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 426307 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 596456 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 626702 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 403677 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 696875 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 67596 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 30082 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 662932 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 28742 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 926850 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 391550 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 101729 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 810457 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 598506 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 61208 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 255036 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 167826 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 501643 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 184467 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 805881 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 929221 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 170190 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 343573 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 109164 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 337455 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 96011 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 386809 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 381146 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 600435 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 364926 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 385025 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 264813 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 864041 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 911065 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 262043 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 841714 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 293421 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 408038 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 39915 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 778586 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 379374 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 640876 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 160074 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 335872 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 741724 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 299763 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 20624 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 492623 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 54973 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 137020 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 113807 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 146218 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 154141 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 75736 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 11285 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 41112 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 45664 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 97377 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 158789 | - | - |
Agilent Whole Rat Genome Microarray 4x44K v3 | A_44_P182402 | - | - |
Agilent Whole Rat Genome Microarray 4x44K v3 | A_64_P158148 | - | - |
Agilent Whole Rat Genome Microarray 4x44K v3 | A_44_P182402 | - | - |
Agilent Whole Rat Genome Microarray 4x44K v3 | A_64_P158148 | - | - |
Agilent SurePrint G3 Rat GE 8x60K Microarray | A_64_P158148 | - | - |
Agilent SurePrint G3 Rat GE 8x60K Microarray | A_44_P182402 | - | - |
SurePrint G3 Rat Gene Expression Microarray G4131A | A_44_P295018 | - | - |
SurePrint G3 Rat Gene Expression Microarray G4131A | A_44_P182402 | - | - |
Agilent Rat Genome CGH Microarray 105A (G4436A) | 63484 | - | - |
Agilent Rat Genome CGH Microarray 105A (G4436A) | 98850 | - | - |
Agilent Rat Genome CGH Microarray 105A (G4436A) | 15795 | - | - |
Agilent Rat Genome CGH Microarray 105A (G4436A) | 58127 | - | - |
Agilent Rat Genome CGH Microarray 105A (G4436A) | 42225 | - | - |
Agilent Rat Genome CGH Microarray 105A (G4436A) | 34520 | - | - |
Agilent Rat Genome CGH Microarray 105A (G4436A) | 65958 | - | - |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Affymetrix Rat Gene 1.0 ST Array | 10717820 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10717815 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10717823 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10717827 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10717817 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10717814 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10717816 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10717819 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10717818 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10717824 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10717826 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10717821 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10717828 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10717822 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10717825 | - | - |
Affymetrix Rat Gene 1.1 ST Array | 10717813 | - | - |
Rat Genome 230 2.0 Array | 1389368_PM_at | - | - |
Rat Genome 230 2.0 Array | 1389368_at | - | - |
Affymetrix HT Rat Focus Array | 1389368_PM_at | - | - |
Affymetrix HT Rat Focus Array | 1389368_at | - | - |
Affymetrix Rat Clariom S Assay | PSR0100034999.rn.1 | TC0100004388.rn.1 | RTA TC |
Affymetrix Rat Clariom S Assay | PSR0100035000.rn.1 | TC0100004388.rn.1 | RTA TC |
Rat Expression Set 230 | 1389368_PM_at | - | - |
Rat Expression Set 230 | 1389368_at | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10717813 | - | - |
Rat Genome U34 Set | rc_AA893357_at | - | - |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
ABI Rat Genome Survey Microarray | 21951010 | - | - |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Illumina ratRef-12 v1.0 expression beadchip | ILMN_1376363 | - | - |
