Assay Details
Gene Symbol: Cask | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_022184.1 | NP_071520.1 | 15 - 16 | 1857 | 97 | |
GenBank mRNA | U47110.1 | - | 15 - 16 | 1857 | 97 |
Gene Symbol: LOC100910506 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_006256650.1 | XP_006256712.1 | 2 - 3 | 263 | 97 | |
XM_006256651.1 | XP_006256713.1 | 2 - 3 | 263 | 97 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
