Assay Details
Gene Symbol: Uxt | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001006982.1 | NP_001006983.1 | 4 - 5 | 356 | 88 | |
XM_006256611.2 | XP_006256673.1 | 5 - 6 | 413 | 88 | ||
GenBank mRNA | BC082752.1 | - | 4 - 5 | 356 | 88 | |
FQ210799.1 | - | 5 - 6 | 401 | 88 | ||
FQ221998.1 | - | 5 - 6 | 88 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
