Assay Details
Gene Symbol: Uxt | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_006256611.2 | XP_006256673.1 | 1 - 2 | 55 | 69 | |
GenBank mRNA | FQ210799.1 | - | 1 - 2 | 43 | 69 | |
FQ221998.1 | - | 1 - 2 | 46 | 69 |

More Information
Additional Information:
This assay is in the 5'UTR region of the XM_006256611.2 transcripts.
Set Membership: |
![]() ![]() ![]() ![]() |
Gene Ontology Categories:
