Assay Details
Gene Symbol: Slc9a7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001108242.1 | NP_001101712.1 | 6 - 7 | 1013 | 97 | |
XM_006256636.2 | XP_006256698.1 | 6 - 7 | 1404 | 97 | ||
XM_006256637.2 | XP_006256699.1 | 6 - 7 | 1404 | 97 |

More Information
Updated Mapping Information

Additional Information:
This assay is in the 5'UTR region of the NM_001108242.1 transcripts.
Set Membership: |
![]() ![]() ![]() |
Gene Ontology Categories:
MicroArray Probes:
