Assay Details
Gene Symbol: Timp1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_053819.1 | NP_446271.1 | 5 - 6 | 483 | 79 | |
XM_006256608.2 | XP_006256670.1 | 5 - 6 | 547 | 79 | ||
XM_008773034.1 | XP_008771256.1 | 4 - 5 | 457 | 79 | ||
GenBank mRNA | AF411319.1 | - | 2 - 3 | 233 | 79 | |
AY550026.1 | - | 4 - 5 | 472 | 79 | ||
BC099821.1 | - | 5 - 6 | 481 | 7930800 | 79 | |
FQ219856.1 | - | 5 - 6 | 493 | 79 | ||
FQ221763.1 | - | 5 - 6 | 493 | 79 | ||
FQ222271.1 | - | 5 - 6 | 493 | 79 | ||
FQ222437.1 | - | 5 - 6 | 496 | 79 | ||
FQ222915.1 | - | 5 - 6 | 493 | 79 | ||
FQ228400.1 | - | 5 - 6 | 492 | 79 | ||
FQ230039.1 | - | 5 - 6 | 490 | 79 | ||
L31883.1 | - | 5 - 6 | 483 | 79 | ||
U06179.1 | - | 4 - 5 | 466 | 79 |

More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
