Assay Details
Gene Symbol: Ddx3x | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001108246.1 | NP_001101716.1 | 10 - 11 | 1128 | 126 | |
XM_006256652.2 | XP_006256714.1 | 12 - 13 | 1424 | 126 | ||
XM_006256653.2 | XP_006256715.1 | 12 - 13 | 1421 | 126 | ||
GenBank mRNA | BC085914.1 | - | 11 - 12 | 1469 | 126 | |
FQ211849.1 | - | 12 - 13 | 1403 | 126 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() |
MicroArray Probes:
