Assay Details
Gene Symbol: Ddx3x | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001108246.1 | NP_001101716.1 | 4 - 5 | 364 | 76 | |
XM_006256652.2 | XP_006256714.1 | 4 - 5 | 400 | 76 | ||
XM_006256653.2 | XP_006256715.1 | 4 - 5 | 397 | 76 | ||
GenBank mRNA | BC085914.1 | - | 4 - 5 | 369 | 76 | |
FQ211849.1 | - | 4 - 5 | 379 | 76 |

More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
This assay is in the 5'UTR region of the NM_001108246.1 transcripts.
This assay is in the 5'UTR region of the NM_001108246.1 transcripts.
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
MicroArray Probes:
