Assay Details
Gene Symbol: Srpx | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_022524.1 | NP_071969.1 | 9 - 10 | 1274 | 82 | |
XM_006256683.2 | XP_006256745.1 | 8 - 9 | 1302 | 82 | ||
XM_008773077.1 | XP_008771299.1 | 8 - 9 | 2428 | 82 | ||
GenBank mRNA | BC087639.1 | - | 8 - 9 | 1273 | 82 | |
D78359.1 | - | 9 - 10 | 1274 | 82 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
Agilent
Affymetrix
ABI
Illumina
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Agilent Whole Rat Genome Microarray 4x44K v2 | A_64_P022746 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 856709 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 745395 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 41371 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 329955 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 173903 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 22543 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 935463 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 11687 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 382506 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 627407 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 742318 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 589550 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 378124 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 781898 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 972259 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 933630 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 374820 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 792894 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 130962 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 673029 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 261439 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 716423 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 7956 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 143834 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 625157 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 54070 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 6618 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 777744 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 835214 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 222232 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 901988 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 232395 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 166722 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 55941 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 317261 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 607526 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 235542 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 562753 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 15357 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 808887 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 449254 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 599232 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 324773 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 927308 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 251355 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 863814 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 14183 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 601043 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 527274 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 250768 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 963250 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 250210 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 73529 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 682093 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 309313 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 368927 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 593728 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 344618 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 58522 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 128721 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 151063 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 125273 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 16557 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 13370 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 123377 | - | - |
Agilent Whole Rat Genome Microarray 4x44K v3 | A_64_P022746 | - | - |
Agilent Whole Rat Genome Microarray 4x44K v3 | A_64_P022746 | - | - |
Agilent SurePrint G3 Rat GE 8x60K Microarray | A_64_P022746 | - | - |
SurePrint G3 Rat Gene Expression Microarray G4131A | A_44_P450737 | - | - |
Agilent Rat Genome CGH Microarray 105A (G4436A) | 12771 | - | - |
Agilent Rat Genome CGH Microarray 105A (G4436A) | 68512 | - | - |
Agilent Rat Genome CGH Microarray 105A (G4436A) | 23101 | - | - |
Agilent Rat Genome CGH Microarray 105A (G4436A) | 28193 | - | - |
Agilent Rat Genome CGH Microarray 105A (G4436A) | 50063 | - | - |
Agilent Rat Genome CGH Microarray 105A (G4436A) | 8321 | - | - |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Affymetrix Rat Gene 1.0 ST Array | 10932362 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10932361 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10932360 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10932357 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10932358 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10932353 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10932363 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10932359 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10932352 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10932356 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10932354 | - | - |
Affymetrix Rat Gene 1.1 ST Array | 10932351 | - | - |
Rat Genome 230 2.0 Array | 1368671_at | - | - |
Rat Genome 230 2.0 Array | 1368671_PM_at | - | - |
Affymetrix HT Rat Focus Array | 1368671_at | - | - |
Affymetrix HT Rat Focus Array | 1368671_PM_at | - | - |
Affymetrix Rat Clariom S Assay | PSR0X00009258.rn.1 | TC0X00001465.rn.1 | RTA TC |
Affymetrix Rat Clariom S Assay | PSR0X00009259.rn.1 | TC0X00001465.rn.1 | RTA TC |
Affymetrix Rat Clariom S Assay | PSR0X00009257.rn.1 | TC0X00001465.rn.1 | RTA TC |
Rat Expression Set 230 | 1368671_PM_at | - | - |
Rat Expression Set 230 | 1368671_at | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10932351 | - | - |
Rat Genome U34 Set | D78359_at | - | - |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
ABI Rat Genome Survey Microarray | 21234937 | - | - |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Illumina ratRef-12 v1.0 expression beadchip | ILMN_1373216 | - | - |
RatRef-12 Expression BeadChip | ILMN_66641 | - | - |
