Assay Details
Gene Symbol: Zdhhc1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001039099.1 | NP_001034188.1 | 7 - 8 | 930 | 67 | |
XM_008772485.1 | XP_008770707.1 | 7 - 8 | 2706 | 67 | ||
XM_008772486.1 | XP_008770708.1 | 7 - 8 | 2705 | 67 | ||
GenBank mRNA | AY886520.1 | - | 7 - 8 | 930 | 67 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() |
Gene Ontology Categories:
MicroArray Probes:
