Assay Details
Gene Symbol: Il15 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
GenBank mRNA | BC097340.1 | - | 1 - 2 | 163 | 131 | |
U69272.1 | - | 556 | 131 |

More Information
Additional Information:
This assay is in the 5'UTR region of the U69272.1 transcripts.
Set Membership: |
![]() ![]() ![]() ![]() |
