Assay Details
Gene Symbol: Wwp2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001106184.1 | NP_001099654.1 | 66 | 70 | ||
GenBank mRNA | JN712752.1 | - | 78 | 70 |
Gene Symbol: LOC103690129 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_008772560.1 | XP_008770782.1 | 1 - 2 | 105 | 70 | |
XM_008772561.1 | XP_008770783.1 | 1 - 2 | 101 | 70 |

More Information
Additional Information:
This assay is in the 5'UTR region of the JN712752.1, NM_001106184.1, XM_008772560.1, XM_008772561.1 transcripts.
Set Membership: |
![]() ![]() ![]() ![]() |
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
