Assay Details
Gene Symbol: Acsf3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_008772664.1 | XP_008770886.1 | 5 - 6 | 1182 | 85 | |
XM_008772665.1 | XP_008770887.1 | 4 - 5 | 649 | 85 | ||
XM_574249.6 | XP_574249.4 | 5 - 6 | 1339 | 85 |

More Information
Set Membership: |
![]() ![]() ![]() |
Gene Ontology Categories:
