Assay Details
Gene Symbol: Acsf3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_008772664.1 | XP_008770886.1 | 8 - 9 | 1553 | 81 | |
XM_008772665.1 | XP_008770887.1 | 7 - 8 | 1020 | 81 | ||
XM_574249.6 | XP_574249.4 | 8 - 9 | 1710 | 81 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
