Assay Details
Gene Symbol: Anapc10 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001108445.1 | NP_001101915.1 | 3 - 4 | 373 | 104 | |
XM_006255406.2 | XP_006255468.1 | 3 - 4 | 375 | 104 | ||
XM_008772516.1 | XP_008770738.1 | 3 - 4 | 329 | 104 | ||
GenBank mRNA | BC168746.1 | - | 3 - 4 | 295 | 6920404 | 104 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
