Assay Details
Gene Symbol: Mphosph6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001129881.2 | NP_001123353.1 | 1 - 2 | 70 | 99 | |
GenBank mRNA | BC166883.1 | - | 1 - 2 | 70 | 99 |

More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Gene Ontology Categories:
MicroArray Probes:
