Assay Details
Gene Symbol: Banp | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001106191.1 | NP_001099661.1 | 8 - 9 | 1209 | 76 | |
XM_006255751.2 | XP_006255813.1 | 8 - 9 | 1255 | 76 | ||
XM_006255752.2 | XP_006255814.1 | 8 - 9 | 1254 | 76 | ||
XM_006255753.2 | XP_006255815.1 | 7 - 8 | 1137 | 76 | ||
XM_006255754.2 | XP_006255816.1 | 7 - 8 | 1137 | 76 | ||
XM_006255755.2 | XP_006255817.1 | 8 - 9 | 1254 | 76 | ||
XM_006255756.2 | XP_006255818.1 | 8 - 9 | 1254 | 76 | ||
XM_006255757.2 | XP_006255819.1 | 7 - 8 | 1136 | 76 | ||
XM_008772601.1 | XP_008770823.1 | 8 - 9 | 1476 | 76 | ||
XM_008772602.1 | XP_008770824.1 | 8 - 9 | 1255 | 76 | ||
GenBank mRNA | BC160844.1 | - | 8 - 9 | 1236 | 8361869 | 76 |

More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Gene Ontology Categories:
MicroArray Probes:
