Assay Details
Gene Symbol: Mvd | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_031062.2 | NP_112324.1 | 7 - 8 | 941 | 66 | |
GenBank mRNA | BC081784.1 | - | 7 - 8 | 926 | 7135885 | 66 |
U53706.1 | - | 7 - 8 | 942 | 66 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
