Assay Details
Gene Symbol: Pdpr | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001107430.1 | NP_001100900.1 | 14 - 15 | 2005 | 80 | |
XM_006255596.2 | XP_006255658.2 | 16 - 17 | 2260 | 80 | ||
XM_008772514.1 | XP_008770736.1 | 15 - 16 | 2176 | 80 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() |
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
