Assay Details
Gene Symbol: Pdpr | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001107430.1 | NP_001100900.1 | 16 - 17 | 2275 | 92 | |
XM_006255596.2 | XP_006255658.2 | 18 - 19 | 2530 | 92 | ||
XM_008772514.1 | XP_008770736.1 | 17 - 18 | 2446 | 92 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Gene Ontology Categories:
MicroArray Probes:
Agilent
Affymetrix
ABI
Illumina
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Agilent Whole Rat Genome Microarray 4x44K v2 | A_44_P171064 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 851160 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 586084 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 299876 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 624881 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 469465 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 891130 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 873180 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 140875 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 574488 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 557199 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 901310 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 56660 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 822863 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 419962 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 174411 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 300781 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 890505 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 213905 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 323672 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 315886 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 834458 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 553091 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 339556 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 58782 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 7323 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 15243 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 158710 | - | - |
Agilent Whole Rat Genome Microarray 4x44K v3 | A_44_P171064 | - | - |
Agilent Whole Rat Genome Microarray 4x44K v3 | A_44_P171064 | - | - |
Agilent SurePrint G3 Rat GE 8x60K Microarray | A_44_P171064 | - | - |
SurePrint G3 Rat Gene Expression Microarray G4131A | A_44_P171064 | - | - |
Agilent Rat Genome CGH Microarray 105A (G4436A) | 3922 | - | - |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Affymetrix Rat Gene 1.0 ST Array | 10808066 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10808082 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10808075 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10808069 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10808080 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10808070 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10808071 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10808074 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10808076 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10808081 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10808077 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10808078 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10808072 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10808068 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10808067 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10808079 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10808073 | - | - |
Affymetrix Rat Gene 1.1 ST Array | 10808065 | - | - |
Rat Genome 230 2.0 Array | 1392726_at | - | - |
Rat Genome 230 2.0 Array | 1392726_PM_at | - | - |
Affymetrix HT Rat Focus Array | 1392726_at | - | - |
Affymetrix HT Rat Focus Array | 1392726_PM_at | - | - |
Affymetrix Rat Clariom S Assay | PSR1900011894.rn.1 | TC1900001460.rn.1 | RTA TC |
Affymetrix Rat Clariom S Assay | PSR1900011895.rn.1 | TC1900001460.rn.1 | RTA TC |
Rat Expression Set 230 | 1392726_PM_at | - | - |
Rat Expression Set 230 | 1392726_at | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10808065 | - | - |
Rat Genome U34 Set | rc_AI059453_at | - | - |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
ABI Rat Genome Survey Microarray | 21255908 | - | - |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Illumina ratRef-12 v1.0 expression beadchip | ILMN_1367404 | - | - |
