Assay Details
Gene Symbol: Adat1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001109611.1 | NP_001103081.1 | 4 - 5 | 445 | 62 | |
XM_006255634.2 | XP_006255696.1 | 4 - 5 | 1285 | 62 | ||
XM_006255635.2 | XP_006255697.1 | 4 - 5 | 1411 | 62 | ||
XM_006255636.2 | XP_006255698.1 | 4 - 5 | 1239 | 62 | ||
XM_006255638.1 | XP_006255700.1 | 3 - 4 | 290 | 62 | ||
XM_008772535.1 | XP_008770757.1 | 4 - 5 | 1285 | 62 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() |
Gene Ontology Categories:
MicroArray Probes:
