Assay Details
Gene Symbol: Adat1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001109611.1 | NP_001103081.1 | 5 - 6 | 569 | 105 | |
XM_006255634.2 | XP_006255696.1 | 5 - 6 | 1409 | 105 | ||
XM_006255635.2 | XP_006255697.1 | 5 - 6 | 1535 | 105 | ||
XM_006255636.2 | XP_006255698.1 | 5 - 6 | 1363 | 105 | ||
XM_006255638.1 | XP_006255700.1 | 4 - 5 | 414 | 105 | ||
XM_008772535.1 | XP_008770757.1 | 5 - 6 | 1409 | 105 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() |
Gene Ontology Categories:
MicroArray Probes:
