Assay Details
Gene Symbol: Cenpn | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001008366.1 | NP_001008367.1 | 2 - 3 | 207 | 65 | |
XM_006255686.2 | XP_006255748.1 | 2 - 3 | 321 | 65 | ||
XM_006255687.2 | XP_006255749.1 | 2 - 3 | 239 | 65 | ||
XM_006255691.2 | XP_006255753.1 | 2 - 3 | 323 | 65 | ||
XM_006255692.2 | XP_006255754.1 | 2 - 3 | 203 | 65 | ||
GenBank mRNA | BC085839.1 | - | 2 - 3 | 207 | 7135799 | 65 |

More Information
Additional Information:
This assay is in the 5'UTR region of the BC085839.1, NM_001008366.1, XM_006255686.2, XM_006255687.2, XM_006255691.2, XM_006255692.2 transcripts.
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
