Assay Details
Gene Symbol: Slc7a6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001107424.1 | NP_001100894.1 | 3 - 4 | 830 | 97 | |
XM_006255498.2 | XP_006255560.1 | 4 - 5 | 895 | 97 | ||
XM_006255499.2 | XP_006255561.1 | 4 - 5 | 925 | 97 | ||
XM_006255500.2 | XP_006255562.1 | 4 - 5 | 1043 | 97 | ||
XM_008772500.1 | XP_008770722.1 | 4 - 5 | 929 | 97 | ||
XM_008772501.1 | XP_008770723.1 | 4 - 5 | 1110 | 97 | ||
XM_008772502.1 | XP_008770724.1 | 4 - 5 | 1159 | 97 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() |
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
