Assay Details
Gene Symbol: Pla2g15 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001004277.2 | NP_001004277.1 | 4 - 5 | 528 | 86 | |
GenBank mRNA | AY490816.1 | - | 4 - 5 | 500 | 86 | |
BC098894.1 | - | 4 - 5 | 528 | 7099635 | 86 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
