Assay Details
Gene Symbol: Exoc3l1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001106178.1 | NP_001099648.1 | 3 - 4 | 467 | 80 | |
XM_006255456.2 | XP_006255518.1 | 4 - 5 | 884 | 80 | ||
XM_006255457.2 | XP_006255519.1 | 4 - 5 | 1220 | 80 | ||
XM_006255460.2 | XP_006255522.1 | 3 - 4 | 398 | 80 | ||
XM_006255461.2 | XP_006255523.1 | 3 - 4 | 347 | 80 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() |
Gene Ontology Categories:
MicroArray Probes:
