Assay Details
Gene Symbol: Cog8 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001106182.2 | NP_001099652.1 | 2 - 3 | 613 | 92 | |
GenBank mRNA | BC168966.1 | - | 2 - 3 | 616 | 92 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() |
MicroArray Probes:
