Assay Details
Gene Symbol: Bco1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_053648.3 | NP_446100.2 | 9 - 10 | 1406 | 73 | |
GenBank mRNA | FQ219222.1 | - | 9 - 10 | 1457 | 73 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
