Assay Details
Gene Symbol: Psmb10 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001025637.1 | NP_001020808.1 | 2 - 3 | 257 | 109 | |
GenBank mRNA | BC098835.1 | - | 2 - 3 | 257 | 7378841 | 109 |
FQ226463.1 | - | 2 - 3 | 250 | 109 | ||
FQ228226.1 | - | 2 - 3 | 238 | 109 | ||
FQ229295.1 | - | 2 - 3 | 255 | 109 | ||
FQ229779.1 | - | 310 | 109 | |||
FQ233484.1 | - | 2 - 3 | 267 | 109 | ||
FQ234080.1 | - | 2 - 3 | 259 | 109 | ||
FQ235075.1 | - | 2 - 3 | 240 | 109 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
