Assay Details
Gene Symbol: Psmb10 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001025637.1 | NP_001020808.1 | 3 - 4 | 359 | 89 | |
GenBank mRNA | BC098835.1 | - | 3 - 4 | 359 | 89 | |
FQ226463.1 | - | 3 - 4 | 352 | 89 | ||
FQ228226.1 | - | 3 - 4 | 340 | 89 | ||
FQ229295.1 | - | 3 - 4 | 357 | 89 | ||
FQ229779.1 | - | 412 | 89 | |||
FQ233484.1 | - | 3 - 4 | 369 | 89 | ||
FQ234080.1 | - | 3 - 4 | 361 | 89 | ||
FQ235075.1 | - | 3 - 4 | 342 | 89 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
