Assay Details
Gene Symbol: Psmb10 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001025637.1 | NP_001020808.1 | 6 - 7 | 669 | 76 | |
GenBank mRNA | BC098835.1 | - | 6 - 7 | 669 | 76 | |
FQ217478.1 | - | 2 - 3 | 212 | 76 | ||
FQ226463.1 | - | 6 - 7 | 662 | 76 | ||
FQ228226.1 | - | 6 - 7 | 649 | 76 | ||
FQ229295.1 | - | 6 - 7 | 667 | 76 | ||
FQ233484.1 | - | 6 - 7 | 679 | 76 | ||
FQ234080.1 | - | 6 - 7 | 671 | 76 | ||
FQ235075.1 | - | 6 - 7 | 652 | 76 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
