Assay Details
Gene Symbol: Psmb10 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001025637.1 | NP_001020808.1 | 1 - 2 | 169 | 70 | |
GenBank mRNA | BC098835.1 | - | 1 - 2 | 169 | 70 | |
FQ226463.1 | - | 1 - 2 | 162 | 70 | ||
FQ228226.1 | - | 1 - 2 | 150 | 70 | ||
FQ229295.1 | - | 1 - 2 | 167 | 70 | ||
FQ229779.1 | - | 222 | 70 | |||
FQ233484.1 | - | 1 - 2 | 179 | 70 | ||
FQ234080.1 | - | 1 - 2 | 171 | 70 | ||
FQ235075.1 | - | 1 - 2 | 152 | 70 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
