Assay Details
Gene Symbol: Mlkl | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_008772570.1 | XP_008770792.1 | 4 - 5 | 872 | 70 | |
XM_008772571.1 | XP_008770793.1 | 4 - 5 | 827 | 70 | ||
XM_008772572.1 | XP_008770794.1 | 3 - 4 | 738 | 70 |

More Information
Set Membership: |
![]() ![]() ![]() |
Gene Ontology Categories:
