Assay Details
Gene Symbol: Mlkl | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_008772570.1 | XP_008770792.1 | 7 - 8 | 1295 | 76 | |
XM_008772571.1 | XP_008770793.1 | 7 - 8 | 1250 | 76 | ||
XM_008772572.1 | XP_008770794.1 | 6 - 7 | 1161 | 76 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Gene Ontology Categories:
