Assay Details
Gene Symbol: Rnaseh2a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001013234.1 | NP_001013252.1 | 6 - 7 | 1114 | 122 | |
XM_008772414.1 | XP_008770636.1 | 6 - 7 | 644 | 122 | ||
XM_008772415.1 | XP_008770637.1 | 4 - 5 | 428 | 122 | ||
XM_008772416.1 | XP_008770638.1 | 4 - 5 | 424 | 122 | ||
GenBank mRNA | BC086345.1 | - | 6 - 7 | 644 | 122 | |
BC086539.1 | - | 6 - 7 | 1114 | 122 | ||
FQ227287.1 | - | 4 - 5 | 473 | 122 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
